Skip to Content
Merck
D-(+)-Raffinose pentahydrate analytical standard

D-(+)-Raffinose pentahydrate

D-(+)-Raffinose pentahydrate analytical standard
Synonym(s):
O-α-D-Galactopyranosyl-(1→6)-α-D-glucopyranosyl β-D-fructofuranoside, Melitose, Melitriose
Empirical Formula (Hill Notation):
C18H32O16 · 5H2O
CAS Number:
Molecular Weight:
594.51

Products

Product Category
Biological Source
Color
Formula Weight
Melting Point (°C)
Physical Form
Product Line
Purity
Format
Analyte Suitability
Tissue
Quality Segment
Application
Optical Isomer
Reference Material Type
Special Grade
Technique