
Cyproheptadine Related Compound A

Synonym(s):
5H-Dibenzo[a,d]cycloheptene
Empirical Formula (Hill Notation):
C15H12
CAS Number:
Molecular Weight:
192.26
Products
Brand
Formula Weight
Manufacturer
Physical Form
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2