
Cyclomethicone 6

Synonym(s):
Dodecamethylcyclohexasiloxane, 2,2,4,4,6,6,8,8,10,10,12,12-Dodecamethylcyclohexasiloxane
Empirical Formula (Hill Notation):
C12H36O6Si6
CAS Number:
Molecular Weight:
444.92
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing