
Clenpenterol hydrochloride

Synonym(s):
1-(4-Amino-3,5-dichlorophenyl)-2-(1,1-dimethylpropylamino)ethanol hydrochloride
Empirical Formula (Hill Notation):
C13H20Cl2N2O · HCl
CAS Number:
Molecular Weight:
327.68
Products
Brand
Formula Weight
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing