![C1 Esterase Inhibitor from human plasma aqueous solution, ≥95% (SDS-PAGE)](/deepweb/assets/sigmaaldrich/product/images/332/886/f5268554-014d-4152-9884-99d85e794661/640/f5268554-014d-4152-9884-99d85e794661.jpg)
C1 Esterase Inhibitor
![C1 Esterase Inhibitor from human plasma aqueous solution, ≥95% (SDS-PAGE)](/deepweb/assets/sigmaaldrich/product/images/332/886/f5268554-014d-4152-9884-99d85e794661/640/f5268554-014d-4152-9884-99d85e794661.jpg)
Synonym(s):
Complement C1 esterase inhibitor, Esterase inhibitor C-1 from human plasma
CAS Number:
Products
Product Category
Brand
Biological Source
Gene Alias
Physical Form
Purity
Tissue
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing