
BMP-4 human

Synonym(s):
BMP-2B, DVR4, BMP-4, Bone morphogenetic protein 2B, Bone morphogenetic protein 4
Products
Product Category
Brand
Biological Source
Gene Alias
Physical Form
Purity
Recombinant Host
Quality Segment
Technique
Showing 1-4 of 4
Compare
Product Number
Description
Pricing