
Bicalutamide for system suitability

Synonym(s):
Bicalutamide (CDX), Casodex, Cosudex, N-[4-Cyano-3-(trifluoromethyl)phenyl]-3-[(4-fluorophenyl)sulfonyl]-2-hydroxy-2-methylpropanamide
Empirical Formula (Hill Notation):
C18H14F4N2O4S
CAS Number:
Molecular Weight:
430.37
Products
Formula Weight
Gene Alias
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing