
Benzalkonium chloride (C10)

Synonym(s):
Benzyldimethyldecylammonium chloride, Benzyldecyldimethylammonium chloride
Linear Formula:
CH3(CH2)9N(Cl)(CH3)2CH2C6H5
CAS Number:
Molecular Weight:
311.93
Products
Brand
Formula Weight
Quality Segment
Markush Class
Markush Group
Package Size
API Family
Showing 1-1 of 1
Compare
Product Number
Description
Pricing