Skip to Content
Merck
9-Azabicyclo[3.3.1]nonane N-oxyl 95%

9-Azabicyclo[3.3.1]nonane N-oxyl

9-Azabicyclo[3.3.1]nonane N-oxyl 95%
Synonym(s):
9-Azabicyclo[3.3.1]nonane N-oxyl radical, ABNO
Empirical Formula (Hill Notation):
C8H14NO
CAS Number:
Molecular Weight:
140.20

Products

Product Category
Brand
Sustainability
Feature
Formula Weight
Melting Point (°C)
Physical Form
Purity
Reagent Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing