7-Aminodesacetoxycephalosporanic acid
Synonym(s):
7-ADCA
Empirical Formula (Hill Notation):
C8H10N2O3S
CAS Number:
Molecular Weight:
214.24
Products
Product Category
Brand
Formula Weight
Manufacturer
Physical Form
Activity Spectrum
Format
Quality Segment
Application
Mode of Action
API Family
Reference Material Type
Special Grade