
5α-Tetrahydrocortisol

Synonym(s):
5α-Pregnane-3α,11β,17α,21-tetrol-20-one, 3α,11β,17α,21-Tetrahydroxy-5α-pregnan-20-one, 3α,5α-Tetrahydrocortisol, Allopregnane-3α,11β,17α,21-tetrol-20-one, Allotetrahydrocortisol, Kendall’s compound C, Reichstein’s substance C
Empirical Formula (Hill Notation):
C21H34O5
CAS Number:
Molecular Weight:
366.49
Products
Brand
Color
Formula Weight
Physical Form
Chemical Composition
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing