
4-(Bromomethyl)-2-chloro-1-fluorobenzene

Synonym(s):
3-Chloro-4-fluorobenzyl bromide
Empirical Formula (Hill Notation):
C7H5BrClF
CAS Number:
Molecular Weight:
223.47
Products
Product Category
Brand
Formula Weight
Physical Form
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing