
2-(1H-Pyrrol-1-yl)ethanamine

Empirical Formula (Hill Notation):
C6H10N2
CAS Number:
Molecular Weight:
110.16
Products
Product Category
Brand
Formula Weight
Physical Form
Quality Segment
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing