Skip to Content
Merck
1,2,3,4-Tetraphenyl-1,3-cyclopentadiene 95%

1,2,3,4-Tetraphenyl-1,3-cyclopentadiene

1,2,3,4-Tetraphenyl-1,3-cyclopentadiene 95%
Empirical Formula (Hill Notation):
C29H22
CAS Number:
Molecular Weight:
370.48
EC Number:

Products

Product Category
Formula Weight
Melting Point (°C)
Physical Form
Purity
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing