Skip to Content
Merck
All Photos(1)

Documents

P5906

Millipore

Protein A (extracellular)–Agarose from Staphylococcus aureus

lyophilized powder

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
41106500
NACRES:
NA.56

biological source

Staphylococcus aureus

form

lyophilized powder

extent of labeling

~2 mg per mL

matrix

Cross-linked 4% beaded agarose

matrix activation

cyanogen bromide

matrix attachment

amino

matrix spacer

1 atom

capacity

≥20 mg/mL (Human IgG)

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Application

Protein A-agarose is used for affinity chromatography, antibody purification and characterization, and protein A, G and L resins. Protein A-agarose has been used to study the effects of protein A immunoadsorption in patients with chronic dilated cardiomyopathy as well as to study multiple sclerosis and gastric cancer.

Quantity

Swelling: 1 g swells to approx. 4 ml.

Physical form

Lyophilized powder stabilized with lactose

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

D Mahadevan et al.
Oncogene, 26(27), 3909-3919 (2007-02-28)
KIT or alpha-platelet-derived growth factor receptor (alpha-PDGFR) activating mutations are the pathogenic mechanisms that characterize gastrointestinal stromal tumors (GIST). Despite excellent responses to imatinib mesylate (IM), patients are relapsing. We developed an IM-resistant GIST cell line (GIST-R) from the IM-sensitive
I Politis et al.
Journal of dairy science, 78(3), 484-490 (1995-03-01)
The effect was examined of individual caseins on the rate of plasminogen activation by bovine urokinase-type and tissue-type plasminogen activators. All individual caseins (alpha-CN, beta-CN, and kappa-CN) enhanced the activity of both types of plasminogen activators. Optimal concentrations for alpha-CN
Gina-Marie Barletta et al.
The Journal of biological chemistry, 278(21), 19266-19271 (2003-03-21)
Glomerular visceral epithelial cells (podocytes) appear to play a central role in maintaining the selective filtration barrier of the renal glomerulus. While the immunoglobulin superfamily member Nephrin was proposed to act as a cell adhesion molecule at the podocyte intercellular
Tao Wang et al.
iScience, 25(1), 103676-103676 (2022-01-18)
The Wiskott-Aldrich syndrome protein and SCAR homolog (WASH), an actin nucleation-promoting factor, is present in the nucleus where it regulates gene transcription and maintains nuclear organization. Here, we show that WASH interacts with core non-homologous end-joining (NHEJ) factors including Ku70/Ku80
K Klaamas et al.
Neoplasma, 55(2), 143-150 (2008-02-02)
All human immunoglobulins are glycosylated. The changes in IgG glycosylation are associated with autoimmune disorders and pregnancy. Little is known about IgG glycosylation in patients with cancer. A lectin enzyme-linked immunosorbent assay (LELISA) based method was developed for measuring the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service