Skip to Content
Merck
All Photos(2)

Key Documents

N1884

Sigma-Aldrich

4-Nitrophenyl β-D-maltoside

β-maltosidase substrate

Synonym(s):

4-Nitrophenyl-beta-D-maltopyranoside

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H25NO13
CAS Number:
Molecular Weight:
463.39
MDL number:
UNSPSC Code:
12352204
PubChem Substance ID:
NACRES:
NA.28

Quality Level

Assay

≥98% (TLC)

form

powder

solubility

water: 19.60-20.40 mg/mL, clear, colorless to light yellow

storage temp.

−20°C

SMILES string

OCC1OC(OC2C(O)C(O)C(OC2CO)Oc3ccc(cc3)N(=O)=O)C(O)C(O)C1O

InChI

1S/C18H25NO13/c20-5-9-11(22)12(23)14(25)18(30-9)32-16-10(6-21)31-17(15(26)13(16)24)29-8-3-1-7(2-4-8)19(27)28/h1-4,9-18,20-26H,5-6H2

InChI key

IAYJZWFYUSNIPN-UHFFFAOYSA-N

General description

4-Nitrophenyl β-D-maltoside is a substrate for amyloglucosidase enzyme and releases chromogenic end product p-nitrophenol, which is measured colorimetrically at 410 nm.

Application

4-Nitrophenyl β-D-maltoside has been used as a substrate for amyloglucosidase enzyme activity.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

G Telford et al.
Medical and veterinary entomology, 26(3), 291-299 (2012-05-25)
The wound-healing maggot, Lucilia sericata Meigen (Diptera: Calliphoridae), degrades extracellular matrix components by releasing enzymes. The purpose of this study was to investigate the glycosylation profiles of wound slough/eschar from chronic venous leg ulcers and the complementary presence of glycosidase
Enzymatic preparation of mushroom dietary fibre: A comparison between analytical and industrial enzymes
Wong, K.H, et al
Food Chemistry, 115(3), 795-800 (2009)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service