Skip to Content
Merck
All Photos(2)

Key Documents

AV46106

Sigma-Aldrich

Anti-DFNA5 antibody produced in rabbit

IgG fraction of antiserum

Synonym(s):

Anti-Deafness, autosomal dominant 5, Anti-ICERE-1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

47 kDa

species reactivity

human

concentration

0.5 mg - 1 mg/mL

technique(s)

immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... DFNA5(1687)

Related Categories

Immunogen

Synthetic peptide directed towards the C terminal region of human DFNA5

Application

Anti-DFNA5 antibody produced in rabbit is suitable for western blotting at a concentration of 1.25μg/ml and for immunohistochemistry at a concentration of 4-8μg/ml.

Biochem/physiol Actions

DFNA5 gene encodes a protein non-syndromic hearing impairment protein 5 also referred to as autosomal dominant 5 that expresses in foetal cochlea. It plays a pivotal role in apoptosis and cell survival. DFNA5 cooperates with TP53 and facilitates the p53-regulated cellular response to genotoxic stress.

Sequence

Synthetic peptide located within the following region: AALLGTCCKLQIIPTLCHLLRALSDDGVSDLEDPTLTPLKDTERFGIVQR

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yoshiko Masuda et al.
Journal of human genetics, 51(8), 652-664 (2006-08-10)
The tumor suppressor p53 plays a crucial role in the cellular response to DNA damage by transcriptional activation of numerous downstream genes. Although a considerable number of p53 target genes have been reported, the precise mechanism of p53-regulated tumor suppression
Ken Op de Beeck et al.
European journal of human genetics : EJHG, 19(9), 965-973 (2011-04-28)
DFNA5 was first identified as a gene causing autosomal dominant hearing loss (HL). Different mutations have been found, all exerting a highly specific gain-of-function effect, in which skipping of exon 8 causes the HL. Later reports revealed the involvement of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service