Skip to Content
Merck
All Photos(1)

Key Documents

Y0000422

Salmeterol xinafoate

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

(±) 4-Hydroxy-a1-[[[6-(4-phenylbutoxy)hexyl]amino]m-ethyl]-1,3-benzenedimethanol xinafoate, GR 33343X xinafoate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C25H37NO4 · C11H8O3
CAS Number:
Molecular Weight:
603.75
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

salmeterol

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

OC(=O)c1ccc2ccccc2c1O.OCc3cc(ccc3O)C(O)CNCCCCCCOCCCCc4ccccc4

InChI

1S/C25H37NO4.C11H8O3/c27-20-23-18-22(13-14-24(23)28)25(29)19-26-15-7-1-2-8-16-30-17-9-6-12-21-10-4-3-5-11-21;12-10-8-4-2-1-3-7(8)5-6-9(10)11(13)14/h3-5,10-11,13-14,18,25-29H,1-2,6-9,12,15-17,19-20H2;1-6,12H,(H,13,14)

InChI key

XTZNCVSCVHTPAI-UHFFFAOYSA-N

Gene Information

human ... ADRB2(154)

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Salmeterol xinafoate EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Biochem/physiol Actions

β2-adrenoceptor agonist.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

R N Brogden et al.
Allergologia et immunopathologia, 20(2), 72-84 (1992-03-01)
Salmeterol xinafoate, like salbutamol (albuterol), is a saligenin derivative, and a selective beta 2-adrenoceptor agonist. It produces bronchodilation for at least 12 hours following inhalation of a single 50 micrograms dose. Salmeterol is intended for regular twice-daily treatment of reversible
A Jouyban-Gharamaleki et al.
International journal of pharmaceutics, 216(1-2), 33-41 (2001-03-29)
The mole fraction solubility of salmeterol xinafoate was determined in various concentrations of dioxane in aqueous binary mixture. Maximum solubility was observed in 90% v/v dioxane and solubility parameter of the solute was estimated from solubility peak equal to 24.99
R N Brogden et al.
Drugs, 42(5), 895-912 (1991-11-01)
Salmeterol xinafoate, like salbutamol (albuterol), is a saligenin derivative, and a selective beta 2-adrenoceptor agonist. It produces bronchodilation for at least 12 hours following inhalation of a single 50 micrograms dose. Salmeterol is intended for regular twice-daily treatment of reversible

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service