Skip to Content
Merck
All Photos(2)

Documents

850445C

Avanti

14:0-16:0 PC

1-myristoyl-2-palmitoyl-sn-glycero-3-phosphocholine, chloroform

Synonym(s):

1-tetradecanoyl-2-hexadecanoyl-sn-glycero-3-phosphocholine; MPPC; PC(14:0/16:0)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C38H76NO8P
CAS Number:
Molecular Weight:
705.99
UNSPSC Code:
51191904
NACRES:
NA.25

Assay

>99% (TLC)

form

liquid

packaging

pkg of 1 × 2.5 mL (850445C-25mg)
pkg of 1 × 20 mL (850445C-500mg)

manufacturer/tradename

Avanti Research - A Croda Brand 850445C

concentration

10 mg/mL (850445C-25mg)
25 mg/mL (850445C-500mg)

lipid type

cardiolipins
phospholipids

shipped in

dry ice

storage temp.

−20°C

InChI

1S/C38H76NO8P/c1-6-8-10-12-14-16-18-19-21-23-25-27-29-31-38(41)47-36(35-46-48(42,43)45-33-32-39(3,4)5)34-44-37(40)30-28-26-24-22-20-17-15-13-11-9-7-2/h36H,6-35H2,1-5H3/t36-/m1/s1

InChI key

RFVFQQWKPSOBED-PSXMRANNSA-N

General description

Phosphatidylcholine (PC) is dietary source of choline. 14:0-16:0 PC (1-myristoyl-2-palmitoyl-sn-glycero-3-phosphocholine) is a water-soluble lysolipid. 14:0-16:0 PC is a symmetrical phosphatidylcholine.

Application

14:0-16:0 PC (1-myristoyl-2-palmitoyl-sn-glycero-3-phosphocholine) has been used to generate Wnt3a liposomes. It has also been used as a calibration standard for lipidomic analysis by liquid chromatography–tandem mass spectrometry (LC-MS). It may be used in the bilayer membrane to study its miscibility with d-erythro-N-palmitoyl-sphingomyelin (16:0-SM) using differential scanning calorimetry (DSC).

Biochem/physiol Actions

Phosphatidylcholine (PC) is involved in the fusion and transport of membranes. It is known to play a vital role in endocytosis and the catalytic activity of enzymes.

Packaging

30 mL Amber Narrow Mouth Glass Bottle with Screw Cap (850445C-500mg)
5 mL Clear Glass Sealed Ampule (850445C-25mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 Oral - STOT SE 3

Target Organs

Liver,Kidney, Respiratory system

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jeffrey K Mills et al.
Biochimica et biophysica acta, 1716(2), 77-96 (2005-10-12)
The enhanced permeability of lipid bilayer membranes at their gel-to-liquid phase transition has been explained using a "bilayer lipid heterogeneity" model, postulating leaky interfacial regions between still solid and melting liquid phases. The addition of lysolipid to dipalmitoylphosphatidylcholine bilayers dramatically
Bohdana Térová et al.
Biochimica et biophysica acta, 1667(2), 182-189 (2004-12-08)
In this study we have used differential scanning calorimetry (DSC) to study the miscibility of different saturated phosphatidylcholines (PCs) with D-erythro-N-palmitoyl-sphingomyelin (16:0-SM). Information about the miscibility was obtained by observing the thermotropic phase behavior of binary mixtures of saturated PCs
Li-Juan Zhao et al.
Journal of ethnopharmacology, 260, 112943-112943 (2020-05-19)
Arenobufagin (ArBu) is an important anti-tumor ingredient of Chan'su which has long been used as traditional Chinese medicine in clinic for tumor therapy in China. The purpose of our study is to investigate the lipid homeostasis regulation effects of ArBu
Ségolène Hescot et al.
Oncotarget, 8(66), 109924-109940 (2018-01-05)
Mitotane (o,p'DDD), the most effective drug in adrenocortical carcinoma, concentrates into the mitochondria and impacts mitochondrial functions. To address the molecular mechanisms of mitotane action and to identify its potential target, metabolomic and lipidomic approaches as well as imaging analyses
Nathan T Morrell et al.
PloS one, 3(8), e2930-e2930 (2008-08-14)
Wnt signals exercise strong cell-biological and regenerative effects of considerable therapeutic value. There are, however, no specific Wnt agonists and no method for in vivo delivery of purified Wnt proteins. Wnts contain lipid adducts that are required for activity and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service