Key Documents
Z405574
Sigma-Aldrich
J. Young high-vacuum PTFE valve, straight through
stopcock size 8 mm, side arm O.D. 11 mm
About This Item
UNSPSC Code:
40141600
NACRES:
NB.79
manufacturer/tradename
J. Young
side arm O.D.
11 mm
stopcock size
8 mm
General description
The J. Young high-vacuum PTFE valve helps in controlling the atmosphere when using air-sensitive materials.
Features:
  • PTFE piston with unique PTFE-sheathed elastomer O-ring seals
  • Grease-free glass-to-PTFE seals
  • High-vacuum to 10-6 torr