T74802
2,4,6-Trimethylbenzoic acid
99%
Synonym(s):
2-Mesitylenecarboxylic acid
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
(CH3)3C6H2CO2H
CAS Number:
Molecular Weight:
164.20
Beilstein:
1866187
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Quality Level
Assay
99%
mp
152-155 °C (lit.)
SMILES string
Cc1cc(C)c(c(C)c1)C(O)=O
InChI
1S/C10H12O2/c1-6-4-7(2)9(10(11)12)8(3)5-6/h4-5H,1-3H3,(H,11,12)
InChI key
FFFIRKXTFQCCKJ-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
[Kinetics of the interaction of a phosphorylating substrate analog with the Ca-ATPase of myosin and heavy meromyosin].
V M Kodentsova et al.
Nauchnye doklady vysshei shkoly. Biologicheskie nauki, (1)(1), 17-19 (1982-01-01)
I A Kozlov et al.
The Biochemical journal, 192(2), 483-488 (1980-11-15)
The alpha-subunits of factor-F1 ATPase are removed by extraction of submitochondrial particles with 1.75 M-LiCl, with the consequent loss of ATPase activity. ATPase activity is reconstituted by incubation of LiCl-extracted particles with purified alpha-subunits, and the reconstituted ATPase activity is
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service