Skip to Content
Merck
All Photos(1)

Key Documents

936014

Sigma-Aldrich

Sodium bis(trifluoromethanesulfonyl)imide solution

greener alternative

in ethylene carbonate, dimethylcarbonate, and ethylmethylcarbonate, 1.0 M NaTFSI in EC/DMC/EMC=1:1:1 (v/v/v), battery grade

Synonym(s):

1.0 M NaTFSI in EC/DMC/EMC electrolyte

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352600

grade

battery grade

Quality Level

form

solution

greener alternative product characteristics

Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

concentration

0.95-1.10 M (NaPF6)

impurities

≤100 ppm H2O

application(s)

battery manufacturing

greener alternative category

Looking for similar products? Visit Product Comparison Guide

General description

Battery grade sodium bis(trifluoromethanesulfonyl)imide solution in ethylene carbonate, dimethylcarbonate, and ethylmethylcarbonate is a clear solution composed of a 1:1:1 volume ratio of three organic solvents - ethylene carbonate (EC), dimethyl carbonate (DMC), and ethyl methyl carbonate (EMC) - and 1.0 M concentration of sodium bis(trifluoromethanesulfonyl)imide (NaTFSI) as the electrolyte salt.
We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for energy efficiency. Click here for more information.

Application

The main application of 1.0 M NaTFSI in EC/DMC/EMC=1:1:1 (v/v/v), battery grade is as a electrolyte for sodium-ion batteries. We offer the superior electrolyte with controlled impurities, including water, acid, and trace metals content. Ethylene carbonate (EC) is known to form a stable, conductive solid electrolyte interphase (SEI) with many cathode and anode materials, including hard carbon and sodium metal oxides Ethyl methyl carbonate (EMC) and dimethyl carbonate (DMC) have lower viscosities and melting points than EC, and when mixed with EC result in an electrolyte with a good balance between desirable electrochemical properties, high dielectric constant, and low viscosity.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Corr. 1B - STOT RE 2 Oral

Target Organs

Kidney

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

In situ synthesis of hierarchical poly(ionic liquid)-based solid electrolytes for high-safety lithium-ion and sodium-ion batteries
Zhou D, et al.
Nano Energy, 33, 45-54 (2017)
Recent progress and perspective on electrolytes for sodium/potassium-based devices
Pan Q, et al.
Energy Storage Materials, 31, 328-343 (2020)

Articles

Discover the role of electrolytes in sodium-ion batteries, to enhance performance, safety, and sustainability in energy storage solutions.

Discover the role of electrolytes in sodium-ion batteries, to enhance performance, safety, and sustainability in energy storage solutions.

Discover the role of electrolytes in sodium-ion batteries, to enhance performance, safety, and sustainability in energy storage solutions.

Discover the role of electrolytes in sodium-ion batteries, to enhance performance, safety, and sustainability in energy storage solutions.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service