Skip to Content
Merck
All Photos(1)

Key Documents

769908

Sigma-Aldrich

Resomer® LR 706 S, Poly(L-lactide-co-D,L-lactide)

ester terminated, L-Lactide: D,L-Lactide 70:30

Synonym(s):

Resomer® LR 706 S, P(L/DL)LA

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
12162002
NACRES:
NA.23

Quality Level

form

solid

feed ratio

lactide:lactide 70:30

viscosity

3.3-4.2 dL/g

mp

102-111 °C

transition temp

Tg 56-62 °C

storage temp.

2-8°C

Legal Information

RESOMER is a registered trademark of Evonik Rohm GmbH

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service