Skip to Content
Merck
All Photos(1)

Key Documents

712795

Sigma-Aldrich

2-Azido-2-deoxy-D-glucose

≥97.0% (HPLC)

Synonym(s):

2-Azido-2-deoxyglucose, Glycosyl azide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H11N3O5
CAS Number:
Molecular Weight:
205.17
Beilstein:
2216646
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.22

Assay

≥97.0% (HPLC)

form

powder

optical activity

[α]/D 22.5±1.0°, c = 1 in H2O

reaction suitability

reaction type: click chemistry

storage temp.

2-8°C

SMILES string

OC[C@H]1OC(O)[C@H](N=[N+]=[N-])[C@@H](O)[C@@H]1O

InChI

1S/C6H11N3O5/c7-9-8-3-5(12)4(11)2(1-10)14-6(3)13/h2-6,10-13H,1H2/t2-,3-,4-,5-,6?/m1/s1

InChI key

URARQBMUQIRZQO-IVMDWMLBSA-N

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service