Skip to Content
Merck
All Photos(1)

Key Documents

567191

Sigma-Aldrich

2,2,3,3-Tetrafluoro-1,4-butanediol

Synonym(s):

2,2,3,3-Tetrafluorobutanediol, NSC 95113, Tetrafluoro-1,4-butanediol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOCH2CF2CF2CH2OH
CAS Number:
Molecular Weight:
162.08
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

form

solid

bp

110-112 °C/13 mmHg (lit.)

mp

77-82 °C (lit.)

SMILES string

OCC(F)(F)C(F)(F)CO

InChI

1S/C4H6F4O2/c5-3(6,1-9)4(7,8)2-10/h9-10H,1-2H2

InChI key

CDZXJJOGDCLNKX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

2,2,3,3-Tetrafluoro-1,4-butanediol may be used in the synthesis of the following:
  • segmented polyurethanes(SPU 001-002)
  • PFPE(perfluoro-polyether)modified segmented polyurethanes (SPU 003-006)
  • 2,2,3,3-tetrafluorobutane-1,4-diyl dicarbamate
  • 2,2,3,3-tetrafluorobutane-1,4-diyl dinitrate
  • 2,2,3,3-tetrafluoro-4-((trimethylsilyl)methoxy)butanol
Reactant for:
Enzymic synthesis of silicone fluorinated aliphatic polyester amides

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

"Comparison of Photocyclization Reactions of Fluoro-vs Nonfluoro-Substituted Polymethyleneoxy Donor Linked Phthalimides"
Park JH, et al.
Bulletin of the Korean Chemical Society,, 34(4), 1108-1114 (2013)
"Synthesis of nitrate ester and nitramine derivatives of polyfluoro alkyl compounds for high energy materials"
Srinivas D and Ghule.DV
Royal Society of Chemistry Advances, 6(10), 7712-7716 (2016)
Piotr Król et al.
Colloid and polymer science, 290(10), 879-893 (2012-06-19)
The polarity of polyurethane coats was studied on the basis of the goniometric method for determination of wetting angle values, on the basis of calculated surface free energy (SFE) values by the van Oss-Good and Owens-Wendt methods, and on the
"Synthesis and surface properties of environmentally responsive segmented polyurethanes"
Vaidya A and Chaudhury.KM
Journal of Colloid and Interface Science, 249(1), 235-245 (2002)
Piotr Król et al.
Colloid and polymer science, 289(15-16), 1757-1767 (2011-12-02)
WAXS, DSC and AFM methods were employed to compare phase structures of the coatings obtained from waterborne polyurethane cationomers which had been synthesised in the reaction of some diisocyanates (MDI, IPDI, TDI and HDI) with polyoxyethylene glycols (M = 600 and 2,000)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service