Skip to Content
Merck
All Photos(1)

Key Documents

253626

Sigma-Aldrich

16-Doxyl-stearic acid methyl ester

Synonym(s):

2-Ethyl-2-(15-methoxy-15-oxopentadecyl)-4,4-dimethyl-3-oxazolidinyloxy, free radical

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H44NO4
CAS Number:
Molecular Weight:
398.60
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

solubility

methanol: soluble 5%, clear to slightly hazy, yellow

storage temp.

2-8°C

SMILES string

CCC1(CCCCCCCCCCCCCCC(=O)OC)OCC(C)(C)N1[O]

InChI

1S/C23H44NO4/c1-5-23(24(26)22(2,3)20-28-23)19-17-15-13-11-9-7-6-8-10-12-14-16-18-21(25)27-4/h5-20H2,1-4H3

InChI key

XVQIOHOXFFTSLD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

16-Doxyl-stearic acid methyl ester has been employed as spin label:
  • to investigate the changes within the lipid double layer of liposomal membranes
  • to measure the bacterial membrane fluidity by electron spin resonance
Spin label reagent useful for the study of membrane fluidity by electron spin resonance spectrophotometry.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dariusz Man et al.
Journal of biological inorganic chemistry : JBIC : a publication of the Society of Biological Inorganic Chemistry, 16(1), 173-181 (2010-10-22)
Liposomes characterized by membranes featuring diverse fluidity (liquid-crystalline and/or gel phase), prepared from egg yolk lecithin (EYL) and dipalmitoylphosphatidylcholine (DPPC), were doped with selected metalloporphyrins and the time-related structural and dynamic changes within the lipid double layer were investigated. Porphyrin
T Koga et al.
The Journal of antimicrobial chemotherapy, 42(2), 133-140 (1998-09-17)
Plaunotol, a cytoprotective antiulcer agent, has a bactericidal effect against Helicobacter pylori, which may result from interaction of this compound with the bacterial cell membrane. The purpose of the present study was to confirm that plaunotol interacts with the H.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service