MilliporeSigma
  • Comparison between two inhibin B ELISA assays in 46,XY testicular disorders of sex development (DSD) with normal testosterone secretion.
Comparison between two inhibin B ELISA assays in 46,XY testicular disorders of sex development (DSD) with normal testosterone secretion.
Journal of pediatric endocrinology & metabolism : JPEM (2018-01-08)
Guilherme Guaragna-Filho, Antônio Ramos Calixto, Georgette Beatriz De Paula, Laurione Cândido De Oliveira, André Moreno Morcillo, Maricilda Palandi De Mello, Andrea Trevas Maciel-Guerra, Gil Guerra-Junior
RÉSUMÉ
Inhibin B is a hormone produced by the Sertoli cells that can provide important information for the investigation of disorders of sex development (DSD) with 46,XY karyotype. The aim of this study is to compare two enzyme-linked immunosorbent assay (ELISA) assays for dosage of serum inhibin B in patients with 46,XY DSD with normal testosterone secretion. Twenty-nine patients with 46,XY DSD and normal testosterone secretion (partial androgen insensitivity syndrome [PAIS] [n=8]; 5α-reductase deficiency [n=7] and idiopathic 46,XY DSD [n=14]) were included. Molecular analysis of the AR and SRD5A2 genes were performed in all patients and the NR5A1 gene analysis in the idiopathic group. Measurements of inhibin B were performed by two second-generation ELISA assays (Beckman-Coulter and AnshLabs). Assays were compared using the interclass correlation coefficient (ICC) and the Bland-Altman method. ICC was 0.915 [95% confidence interval (CI): 0.828-0.959], however, a discrepancy was observed between trials, which is more evident among higher values when analyzed by the Bland-Altman method. It is recommended to perform the inhibin B measurement always using the same ELISA kit when several evaluations are required for a specific patient.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
Human PLG (Plasminogen) ELISA Kit, for cell culture supernatants, plasma, and serum samples