MilliporeSigma
Palladium-imidazolium carbene catalyzed Mizoroki-Heck coupling with aryl diazonium ions.
Organic letters (2002-06-07)
Merritt B Andrus, Chun Song, Jiuqing Zhang
RÉSUMÉ
[reaction: see text] Catalyst formed from N,N-bis(2,6-diisopropylphenyl)dihydroimidazolium chloride and palladium(II) acetate (2 mol %) was used, without added base, to efficiently produce Heck coupled products with olefins and aryl diazonium tetrafluoroborate substrates. The reactions were performed at room temperature, giving product in 2-4 h with 80-90% yields for isolated materials. Diazonium ions, formed in situ directly from anilines, also couple under these conditions.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
1,3-Bis-(2,6-diisopropylphenyl)imidazolinium chloride, 97%