- A sensitive sandwich-type immunosensor for the detection of galectin-3 based on N-GNRs-Fe-MOFs@AuNPs nanocomposites and a novel AuPt-methylene blue nanorod.
A sensitive sandwich-type immunosensor for the detection of galectin-3 based on N-GNRs-Fe-MOFs@AuNPs nanocomposites and a novel AuPt-methylene blue nanorod.
Biosensors & bioelectronics (2017-11-03)
Zhiyong Tang, Junlin He, Jun Chen, Yazhen Niu, Yilin Zhao, Yuchan Zhang, Chao Yu
PMID29096363
RÉSUMÉ
We are presenting an electrochemical immunosensor for the determination of Galectin-3 (Gal-3), a biomarker of heart failure. A glassy carbon electrode (GCE) was modified with a film of a composite made from the N-doped graphene nanoribbons immobilized Fe-based-Metal-organic frameworks deposited with Au nanoparticles (N-GNRs-Fe-MOFs@AuNPs). Primary antibody against Gal-3 (Gal-3-Ab
MATÉRIAUX