MilliporeSigma
  • Erding Formula in hyperuricaemia treatment: unfolding traditional Chinese herbal compatibility using modern pharmaceutical approaches.
Erding Formula in hyperuricaemia treatment: unfolding traditional Chinese herbal compatibility using modern pharmaceutical approaches.
The Journal of pharmacy and pharmacology (2017-10-24)
Jieyu Zuo, Hongming He, Zhengyun Zuo, Nádia Bou-Chacra, Raimar Löbenberg
RÉSUMÉ
Traditional Chinese herbal formulas are difficult to be understood because of complex compositions and specific therapeutic principles. To better understand herbal compatibility in Traditional Chinese medicine (TCM), this study was conducted to investigate the effects of a Chinese pharmacopoeia-listed formula, Erding Formula (EF) and its constituent herbs for a new indication, hyperuricaemia. A hypoxanthine and potassium oxonate-induced hyperuricemic mouse model, a xylene-induced inflammatory mouse model and an acetic acid-induced pain model were used to test the effects of EF and its constituent herbs. In addition, we investigated whether EF and/or its relevant herbs had an impact on the expression of URAT1 and OAT3 mRNA. The results showed EF and individual herbs had pharmacological effects on selected targets. Only Viola yedoensis Makino (Viola) lowered uric acid levels, while all four herbs had anti-inflammatory and analgesic effects. The EF may lower the uric acid level through inhibiting the expression of URAT1 mRNA and enhancing the expression of OAT3 mRNA. These findings provide pharmacological insights into the effects of EF and individual herbs on UA excretion. This study suggests that Viola is the main herb in EF. This study facilitates better understanding of TCM principles and theories using modern pharmaceutical approaches.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
Oxonic acid potassium salt, 97%