MilliporeSigma
Modification of cellulose foam paper for use as a high-quality biocide disinfectant filter for drinking water.
Carbohydrate polymers (2017-12-20)
Solmaz Heydarifard, Kapila Taneja, Gaurav Bhanjana, Neeraj Dilbaghi, Mousa M Nazhad, Ki-Hyun Kim, Sandeep Kumar
RÉSUMÉ
Development of a foam-formed cellulose filter paper with high wet strength was carried out for application as a drinking water filter. The wet strength and antimicrobial activity of cellulose foam paper against several bacteria species (Bacillus subtilis MTCC 441 (Gram +ve), B. cereus NCDC 240 (Gram +ve), Pseudomonas aeruginosa NCDC 105 (Gram -ve), Klebsiella pneumonia NCDC 138 (Gram -ve), and Escherichia coli MTCC 40 (Gram -ve)) were investigated. The morphology and structure of the cellulose foam paper were characterized using scanning electron microscopy (SEM). The results of our study confirmed that glutaraldehyde solution or 1,2,3,4-butanetetracarboxylic acid (BTCA) added to cellulose foam paper pretreated with cationic polyacrylamide (C-PAM) provided very high and stable wet strength performance together with excellent antimicrobial properties.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
1,2,3,4-Butanetetracarboxylic acid, 99%