MilliporeSigma
  • Chronic and Intermittent Angle Closure Caused by In-The-Bag Capsular Tension Ring and Intraocular Lens Dislocation in Patients With Pseudoexfoliation Syndrome.
Chronic and Intermittent Angle Closure Caused by In-The-Bag Capsular Tension Ring and Intraocular Lens Dislocation in Patients With Pseudoexfoliation Syndrome.
Journal of glaucoma (2017-10-28)
Frank Bochmann, Jörg Strümer
RÉSUMÉ
A subluxation or dislocation of the intraocular lens (IOL) is a well-known complication after cataract surgery in patients with pseudoexfoliation syndrome (PEX). Capsular tension rings (CTR) are frequently used to prevent postoperative complications caused by zonular weakness. Here we present a series of 7 cases, 2 with intermittent and 5 with acute angle closure caused by a combination of PEX-associated zonular weakness and a resulting unstable capsular bag-CTR-IOL complex. We describe the typical clinical course with elevated intraocular pressure, myopic shift, and shallowing of the anterior chamber in 7 patients with a new type of secondary angle closure. The diagnostic challenges and treatment options are discussed in detail. In all cases, the mechanism of angle closure identified by ultrasound biomicroscopy was an anterior dislocation of the peripheral iris by the capsular bag-CTR-IOL complex. The IOL was removed together with the capsular bag and the CTR and replaced by an iris-fixated IOL in all cases. We present a new type of secondary angle closure caused by an anterior dislocated capsular bag-CTR-IOL complex. The cause of both, this atypical presentation and the ineffectiveness of the laser peripheral iridotomy is the large diameter CTR. Therefore CTR implantation might also be the source of late postoperative complications. If a secondary angle closure associated with PEX and unstable capsular bag-CTR-IOL complex is encountered, IOL explantation and replacement by an iris-fixated IOL is a simple and effective treatment option.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
Dorzolamide, ≥98% (HPLC)