MilliporeSigma
Surface functionalization of cellulose with poly(3-hexylthiophene) via novel oxidative polymerization.
Carbohydrate polymers (2017-11-08)
Thien An Phung Hai, Ryuichi Sugimoto
RÉSUMÉ
Surface functionalization of cellulose with poly(3-hexylthiophene) (P3HT) was conducted with FeCl
MATÉRIAUX
Numéro du produit
Marque
Description du produit
Sigma-Aldrich
3-Hexylthiophene, ≥99%