MilliporeSigma
FSTL1 Promotes Metastasis and Chemoresistance in Esophageal Squamous Cell Carcinoma through NFκB-BMP Signaling Cross-talk.
Cancer research (2017-09-09)
Marco Chi-Chung Lau, Kai Yu Ng, Tin Lok Wong, Man Tong, Terence K Lee, Xiao-Yan Ming, Simon Law, Nikki P Lee, Annie L Cheung, Yan-Ru Qin, Kwok Wah Chan, Wen Ning, Xin-Yuan Guan, Stephanie Ma
RÉSUMÉ
Esophageal squamous cell carcinoma (ESCC) has a generally poor prognosis, and molecular markers to improve early detection and predict outcomes are greatly needed. Here, we report that the BMP-binding follistatin-like protein FSTL1 is overexpressed in ESCCs, where it correlates with poor overall survival. Genetic amplification of FSTL1 or chromosome 3q, where it is located, occurred frequently in ESCC, where FSTL1 copy number correlated positively with higher FSTL1 protein expression. Elevating FSTL1 levels by various means was sufficient to drive ESCC cell proliferation, clonogenicity, migration, invasion, self-renewal, and cisplatin resistance