MilliporeSigma
An instrumental approach to combining confocal microspectroscopy and 3D scanning probe nanotomography.
Ultramicroscopy (2017-07-04)
Konstantin E Mochalov, Anton A Chistyakov, Daria O Solovyeva, Alexey V Mezin, Vladimir A Oleinikov, Ivan S Vaskan, Michael Molinari, Igor I Agapov, Igor Nabiev, Anton E Efimov
RÉSUMÉ
In the past decade correlative microscopy, which combines the potentials of different types of high-resolution microscopies with a variety of optical microspectroscopy techniques, has been attracting increasing attention in material science and biological research. One of outstanding solutions in this area is the combination of scanning probe microscopy (SPM), which provides data on not only the topography, but also the spatial distribution of a wide range of physical properties (elasticity, conductivity, etc.), with ultramicrotomy, allowing 3D multiparametric examination of materials. The combination of SPM and ultramicrotomy (scanning probe nanotomography) is very appropriate for characterization of soft multicompound nanostructurized materials, such as polymer matrices and microstructures doped with different types of nanoparticles (magnetic nanoparticles, quantum dots, nanotubes, etc.), and biological materials. A serious problem of this technique is a lack of chemical and optical characterization tools, which may be solved by using optical microspectroscopy. Here, we report the development of an instrumental approach to combining confocal microspectroscopy and 3D scanning probe nanotomography in a single apparatus. This approach retains all the advantages of SPM and upright optical microspectroscopy and allows 3D multiparametric characterization using both techniques. As the first test of the system developed, we have performed correlative characterization of the morphology and the magnetic and fluorescent properties of fluorescent magnetic microspheres doped with a fluorescent dye and magnetic nanoparticles. The results of this study can be used to obtain 3D volume images of a specimen for most high-resolution near-field scanning probe microscopies: SNOM, TERS, AFM-IR, etc. This approach will result in development of unique techniques combining the advantages of SPM (nanoscale morphology and a wide range of physical parameters) and high-resolution optical microspectroscopy (nanoscale chemical mapping and optical properties) and allowing simultaneous 3D measurements.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
Epoxy Embedding Medium kit, embedding resin for electron microscopy