MilliporeSigma
  • Establishment of a novel hepatocyte model that expresses four cytochrome P450 genes stably via mammalian-derived artificial chromosome for pharmacokinetics and toxicity studies.
Establishment of a novel hepatocyte model that expresses four cytochrome P450 genes stably via mammalian-derived artificial chromosome for pharmacokinetics and toxicity studies.
PloS one (2017-10-25)
Daisuke Satoh, Satoru Iwado, Satoshi Abe, Kanako Kazuki, Shinobu Wakuri, Mitsuo Oshimura, Yasuhiro Kazuki
RÉSUMÉ
The utility of HepG2 cells to assess drug metabolism and toxicity induced by chemical compounds is hampered by their low cytochrome P450 (CYP) activities. To overcome this limitation, we established HepG2 cell lines expressing major CYP enzymes involved in drug metabolism (CYP2C9, CYP2C19, CYP2D6, and CYP3A4) and CYP oxidoreductase (POR) using the mammalian-derived artificial chromosome vector. Transchromosomic HepG2 (TC-HepG2) cells expressing four CYPs and POR were used to determine time- and concentration-dependent inhibition and toxicity of several compounds by luminescence detection of CYP-specific substrates and cell viability assays. Gene expression levels of all four CYPs and POR, as well as the CYP activities, were higher in TC-HepG2 clones than in parental HepG2 cells. Additionally, the activity levels of all CYPs were reduced in a concentration-dependent manner by specific CYP inhibitors. Furthermore, preincubation of TC-HepG2 cells with CYP inhibitors known as time-dependent inhibitors (TDI) prior to the addition of CYP-specific substrates determined that CYP inhibition was enhanced in the TDI group than in the non-TDI group. Finally, the IC50 of bioactivable compound aflatoxin B1 was lower in TC-HepG2 cells than in HepG2 cells. In conclusion, the TC-HepG2 cells characterized in the current study are a highly versatile model to evaluate drug-drug interactions and hepatotoxicity in initial screening of candidate drug compounds, which require a high degree of processing capacity and reliability.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
Tienilic Acid, ≥98% (HPLC)