MilliporeSigma
  • Impact of renal function on the underlying pathophysiology of coronary plaque composition in patients with type 2 diabetes mellitus.
Impact of renal function on the underlying pathophysiology of coronary plaque composition in patients with type 2 diabetes mellitus.
Cardiovascular diabetology (2017-10-14)
Kentaro Kakuta, Kaoru Dohi, Miho Miyoshi, Takashi Yamanaka, Masaki Kawamura, Jun Masuda, Tairo Kurita, Toru Ogura, Norikazu Yamada, Yasuhiro Sumida, Masaaki Ito
RÉSUMÉ
Both the progression of diabetic kidney disease and increased glycemic variability play important roles in the pathogenesis of coronary plaque formation via inflammatory pathways in patients with type 2 diabetes mellitus (T2DM). Therefore we evaluated the role of renal function in the contributory effects of blood glucose fluctuations and blood levels of inflammatory cytokine concentrations on the tissue characteristics of coronary plaques in patients with T2DM. We prospectively enrolled 71 T2DM patients (mean age: 68 ± 9, male 79%) with 153 coronary artery lesions. Patients were divided into 2 groups according to their estimated glomerular filtration rate (eGFR) levels: Group 1 (≥ 60 mL/min/1.73 m In Group 1, MAGE correlated with percent lipidic volume (%LV) (r = 0.477, p = 0.002). In this group, stepwise multivariate linear regression analyses showed that only MAGE was independently associated with %LV (β = 0.477, p = 0.002). In contrast, in Group 2, only serum TNF-α correlated with percent fibrotic volume (%FV) (r = - 0.471, p = 0.007), %LV (r = 0.496, p = 0.005) and percent necrotic volume (%NV) (r = 0.426, p = 0.017). In this group, stepwise multivariate linear regression analyses showed that only serum TNF-α was independently associated with each tissue characteristic (%FV β = - 0.471 and p = 0.007, %LV β = 0.496 and p = 0.005, %NV: β = 0.426 and p = 0.017). In T2DM patients, the tissue characteristics of coronary plaques were associated with MAGE in patients with eGFR ≥ 60 mL/min/1.73 m
MATÉRIAUX
Référence du produit
Marque
Description du produit
Isosorbide dinitrate, European Pharmacopoeia (EP) Reference Standard