MilliporeSigma
A lipophilic ionic liquid based on formamidinium cations and TFSI: the electric response and the effect of CO
Physical chemistry chemical physics : PCCP (2017-09-22)
Federico Bertasi, Guinevere A Giffin, Keti Vezzù, Giuseppe Pace, Yaser Abu-Lebdeh, Michel Armand, Vito Di Noto
RÉSUMÉ
This work describes the preparation of the new lipophilic ionic liquid tetraoctyl-formamidinium bis(trifluoromethanesulfonyl) imide (TOFATFSI), which is miscible with lower alkanes. In particular, this work focuses on the electric behaviour of TOFATFSI in the particularly challenging highly apolar environment of supercritical CO
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
Dioctylamine, 97%
Sigma-Aldrich
Formamidine hydrochloride, 97%