MilliporeSigma
Screening of "spice" herbal mixtures: From high-field to low-field proton NMR.
Forensic science international (2017-08-31)
Gaëtan Assemat, Florian Dubocq, Stéphane Balayssac, Catherine Lamoureux, Myriam Malet-Martino, Véronique Gilard
RÉSUMÉ
Forty one samples of herbal spices intended to be introduced into the European market and seized by the French customs were analysed with high-field
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
Dimethyl maleate, 96%