MilliporeSigma
Thiol dioxygenase turnover yields benzothiazole products from 2-mercaptoaniline and O
Archives of biochemistry and biophysics (2017-08-23)
William P Morrow, Sinjinee Sardar, Pawan Thapa, Mohammad S Hossain, Frank W Foss, Brad S Pierce
RÉSUMÉ
Thiol dioxygenases are non-heme mononuclear iron enzymes that catalyze the O
MATÉRIAUX
Numéro du produit
Marque
Description du produit
Sigma-Aldrich
2-Aminothiophenol, 99%
Sigma-Aldrich
2-Methylbenzothiazole, 99%