MilliporeSigma
Ethylene glycol, but not DMSO, could replace glycerol inclusion in soybean lecithin-based extenders in ram sperm cryopreservation.
Animal reproduction science (2016-12-25)
Abouzar Najafi, Hossein Daghigh-Kia, Hossein Vaseghi Dodaran, Mahdieh Mehdipour, Manuel Alvarez-Rodriguez
RÉSUMÉ
The aim of this study was to evaluate the effects of glycerol, ethylene glycol or DMSO in a soybean lecithin extender for freezing ram semen. In this study, 20 ejaculates were collected from four Ghezel rams and diluted with soybean lecithin extender with glycerol (7%), ethylene glycol (3%, 5% and 7%) or DMSO (3%, 5% and 7%). Sperm motility (CASA), membrane integrity (HOS test), viability, total abnormality, mitochondrial activity (Rhodamine 123) and apoptotic features (Annexin V/Propidium iodide) were assessed after thawing. There was no significant difference between glycerol and ethylene glycol at different concentrations (3% and 5%) regarding sperm total and progressive motility, viability, and membrane integrity. The least percentages of mitochondrial functionality were observed in samples frozen with all different DMSO concentrations tested (P<0.05). Moreover, the percentage of post-thawed dead sperm was the greatest for all the DMSO concentrations compared with other groups (P<0.05). Thus, DMSO had an adverse effect on the post thaw ram sperm parameters. In contrast, ethylene glycol could be a desirable substitute of glycerol in the freezing extender, in view of similar results obtained in post-thaw quality of ram semen cryopreserved in a soybean lecithin extender. We propose that glycerol in a soybean lecithin based extender could be replaced by ethylene glycol at 3% or 5% concentrations.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
L-α-phosphatidylcholine, from soybean, Type IV-S, ≥30% (enzymatic)