MilliporeSigma
How Low-Income Subsidy Recipients Respond to Medicare Part D Cost Sharing.
Health services research (2016-06-22)
Bruce Stuart, Franklin B Hendrick, Jing Xu, J Samantha Dougherty
RÉSUMÉ
To determine the magnitude and mechanisms of response to Medicare Part D cost sharing by low-income subsidy (LIS) recipients using oral hypoglycemic agents (OHAs) and statins. Medicare data for a 5 percent random sample of beneficiaries with diabetes enrolled in fee-for-service Part D drug plans in 2008. We evaluated the impact of differences between generic and brand cost sharing rates among cohorts of LIS and non-LIS recipients to determine if wider price spreads increased the generic dispensing rate (GDR) and reduced total drug use and cost. We found little association between cost sharing and aggregate OHA and statin use. In adjusted analyses, non-LIS beneficiaries who paid 46 percent of total OHA costs had 2.5 percent fewer OHA days supply than full benefit dual eligibles who paid just 5 percent of their therapy costs. For statins, the difference in days supply between those facing the lowest and highest cost sharing was 4.6 percent. Higher cost sharing was associated with filling fewer but larger prescriptions for both generics and brands. Higher generic and brand copays had little association with OHA and statin use among LIS recipients. This implies that modest changes in required cost sharing for these medicines would have very little substantive impact on generic dispensing or utilization patterns among LIS recipients and thus would have little effect on total program spending. At the same time, any increases in out-of-pocket costs would be expected to shift costs and place greater financial burden on low-income beneficiaries, particularly those in poor health.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Supelco
Lithium 3,5-diiodosalicylate, analytical standard