MilliporeSigma
Moesin and merlin regulate urokinase receptor-dependent endothelial cell migration, adhesion and angiogenesis.
The international journal of biochemistry & cell biology (2017-05-06)
Bernard Degryse, Mishan Britto, Chun Xu Shan, Robert G Wallace, Keith D Rochfort, Philip M Cummins, Gerardene Meade, Ronan P Murphy
RÉSUMÉ
The glycosyl-phosphatidyl-inositol (GPI)-anchored urokinase receptor (uPAR) has no intracellular domain, but nevertheless initiates signalling through proximal interactions with other membrane receptors including integrins. The relationships between uPAR and ezrin/radixin/moesin (ERM) proteins, moesin and merlin have never been explored. Moesin and merlin are versatile membrane-actin links and regulators of receptors signalling, respectively. We show that uPAR controls moesin and merlin, which propagate uPAR-initiated signals and modulate integrin functions, thereby regulating uPAR activity. uPAR rapidly de-phosphorylates moesin and phosphorylates merlin inactivating both proteins, and enhancing cell migration and angiogenesis. Moesin behaves as a molecular switch turning either on or off uPAR signalling through cycles of de-activation/activation, or sustained activation, respectively. Furthermore, moesin is at the crossroads of uPAR-initiated outside-in and inside-out signalling promoting integrin-dependent cell adhesion suggesting that uPAR also activates integrins distally through moesin. Knocking down merlin expression enhanced cell migration and adhesion through different regulation of fibronectin- and vitronectin-binding integrins.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
MISSION® esiRNA, targeting human MSN