MilliporeSigma
Effects of rearing salinity on expression and function of ion-motive ATPases and ion transport across the gastric caecum of
The Journal of experimental biology (2017-07-01)
Natalie M D'Silva, Marjorie L Patrick, Michael J O'Donnell
RÉSUMÉ
Larvae of