MilliporeSigma
  • Genetic association analysis of the RTK/ERK pathway with aggressive prostate cancer highlights the potential role of CCND2 in disease progression.
Genetic association analysis of the RTK/ERK pathway with aggressive prostate cancer highlights the potential role of CCND2 in disease progression.
Scientific reports (2017-07-05)
Yang Chen, Qin Zhang, Qiuyan Wang, Jie Li, Csilla Sipeky, Jihan Xia, Ping Gao, Yanling Hu, Haiying Zhang, Xiaobo Yang, Haitao Chen, Yonghua Jiang, Yuehong Yang, Ziting Yao, Yinchun Chen, Yong Gao, Aihua Tan, Ming Liao, Johanna Schleutker, Jianfeng Xu, Yinghao Sun, Gong-Hong Wei, Zengnan Mo
RÉSUMÉ
The RTK/ERK signaling pathway has been implicated in prostate cancer progression. However, the genetic relevance of this pathway to aggressive prostate cancer at the SNP level remains undefined. Here we performed a SNP and gene-based association analysis of the RTK/ERK pathway with aggressive prostate cancer in a cohort comprising 956 aggressive and 347 non-aggressive cases. We identified several loci including rs3217869/CCND2 within the pathway shown to be significantly associated with aggressive prostate cancer. Our functional analysis revealed a statistically significant relationship between rs3217869 risk genotype and decreased CCND2 expression levels in a collection of 119 prostate cancer patient samples. Reduced expression of CCND2 promoted cell proliferation and its overexpression inhibited cell growth of prostate cancer. Strikingly, CCND2 downregulation was consistently observed in the advanced prostate cancer in 18 available clinical data sets with a total amount of 1,095 prostate samples. Furthermore, the lower expression levels of CCND2 markedly correlated with prostate tumor progression to high Gleason score and elevated PSA levels, and served as an independent predictor of biochemical relapse and overall survival in a large cohort of prostate cancer patients. Together, we have identified an association of genetic variants and genes in the RTK/ERK pathway with prostate cancer aggressiveness, and highlighted the potential importance of CCND2 in prostate cancer susceptibility and tumor progression to metastasis.
MATÉRIAUX
Numéro du produit
Marque
Description du produit
Roche
Kit de prolifération cellulaire II (XTT), liquid, pkg of 1 kit, suitable for cell analysis, suitable for tissue culture
Sigma-Aldrich
MISSION® esiRNA, targeting human CCND2