MilliporeSigma
The use of radiocobalt as a label improves imaging of EGFR using DOTA-conjugated Affibody molecule.
Scientific reports (2017-07-22)
Javad Garousi, Ken G Andersson, Johan H Dam, Birgitte B Olsen, Bogdan Mitran, Anna Orlova, Jos Buijs, Stefan Ståhl, John Löfblom, Helge Thisgaard, Vladimir Tolmachev
RÉSUMÉ
Several anti-cancer therapies target the epidermal growth factor receptor (EGFR). Radionuclide imaging of EGFR expression in tumours may aid in selection of optimal cancer therapy. The
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
1,4,7,10-Tetraazacyclododecane-1,4,7,10-tetraacetic acid, ≥97.0% (CHN)