MilliporeSigma
Implications for breast cancer treatment from increased autotaxin production in adipose tissue after radiotherapy.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology (2017-05-26)
Guanmin Meng, Xiaoyun Tang, Zelei Yang, Matthew G K Benesch, Alison Marshall, David Murray, Denise G Hemmings, Frank Wuest, Todd P W McMullen, David N Brindley
RÉSUMÉ
We have previously established that adipose tissue adjacent to breast tumors becomes inflamed by tumor-derived cytokines. This stimulates autotaxin (ATX) secretion from adipocytes, whereas breast cancer cells produce insignificant ATX. Lysophosphatidate produced by ATX promotes inflammatory cytokine secretion in a vicious inflammatory cycle, which increases tumor growth and metastasis and decreases response to chemotherapy. We hypothesized that damage to adipose tissue during radiotherapy for breast cancer should promote lysophosphatidic acid (LPA) signaling and further inflammatory signaling, which could potentially protect cancer cells from subsequent fractions of radiation therapy. To test this hypothesis, we exposed rat and human adipose tissue to radiation doses (0.25-5 Gy) that were expected during radiotherapy. This exposure increased mRNA levels for ATX, cyclooxygenase-2, IL-1β, IL-6, IL-10, TNF-α, and LPA
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
DL-Glyceraldehyde 3-phosphate solution, 45-55 mg/mL in H2O
Sigma-Aldrich
CGK733, ≥98% (HPLC), solid