MilliporeSigma
Biochemical characterization of an enantioselective esterase from Brevundimonas sp. LY-2.
Microbial cell factories (2017-06-21)
Jing Zhang, Mengjun Zhao, Die Yu, Jingang Yin, Hao Zhang, Xing Huang
RÉSUMÉ
Lactofen, a member of the diphenylether herbicides, has high activity and is commonly used to control broadleaf weeds. As a post-emergent herbicide, it is directly released to the environment, and easily caused the pollution. This herbicide is degraded in soil mainly by microbial activity, but the functional enzyme involved in the biodegradation of lactofen is still not clear now. A novel esterase gene lacH, involved in the degradation of lactofen, was cloned from the strain Brevundimonas sp. LY-2. The gene contained an open reading frame of 921 bp, and a putative signal peptide at the N-terminal was identified with the most likely cleavage site between Ala 28 and Ala 29. The encoded protein, LacH, could catalyze the hydrolysis of lactofen to form acifluorfen. Phylogenetic analysis showed that LacH belong to family V of bacterial lipolytic enzymes. Biochemical characterization analysis showed that LacH was a neutral esterase with an optimal pH of 7.0 and an optimal temperature of 40 °C toward lactofen. Besides, the activity of LacH was strongly inhibited by Hg Our studies characterized a novel esterase involved in the biodegradation of diphenylether herbicide lactofen. The esterase showed enantioselectivity during lactofen degradation, which revealed the occurrence of enzyme-mediated enantioselective degradation of chiral herbicides.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Supelco
Lactofen, PESTANAL®, analytical standard