MilliporeSigma
Variation in Free Radical Yields from U.S. Marketed Cigarettes.
Chemical research in toxicology (2017-03-09)
Reema Goel, Zachary Bitzer, Samantha M Reilly, Neil Trushin, Jonathan Foulds, Joshua Muscat, Jason Liao, Ryan J Elias, John P Richie
RÉSUMÉ
Free radicals in tobacco smoke are thought to be an important cause of smoking-induced diseases, yet the variation in free radical exposure to smokers from different brands of commercially available cigarettes is unknown. We measured the levels of highly reactive gas-phase and stable particulate-phase radicals in mainstream cigarette smoke by electron paramagnetic resonance (EPR) spectroscopy with and without the spin-trapping agent phenyl-N-tert-butylnitrone (PBN), respectively, in 27 popular US cigarettes and the 3R4F research cigarette, machine-smoked according to the FTC protocol. We find a 12-fold variation in the levels of gas-phase radicals (1.2 to 14 nmol per cigarette) and a 2-fold variation in the amounts of particulate-phase radicals (44 to 96 pmol per cigarette) across the range of cigarette brands. Gas and particulate-phase radicals were highly correlated across brands (ρ = 0.62, p < 0.001). Both radicals were correlated with TPM (gas-phase: ρ = 0.38, p = 0.04; particulate-phase: ρ = 0.44, p = 0.02) and ventilation (gas- and tar-phase: ρ = -0.58, p = 0.001), with ventilation explaining nearly 30% of the variation in radical levels across brands. Overall, our findings of significant brand variation in free radical delivery under standardized machine-smoked conditions suggest that the use of certain brands of cigarettes may be associated with greater levels of oxidative stress in smokers.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
2,2,6,6-Tetramethyl-4-piperidinol, 98%