MilliporeSigma
  • Identification of a Novel Benzimidazole Pyrazolone Scaffold That Inhibits KDM4 Lysine Demethylases and Reduces Proliferation of Prostate Cancer Cells.
Identification of a Novel Benzimidazole Pyrazolone Scaffold That Inhibits KDM4 Lysine Demethylases and Reduces Proliferation of Prostate Cancer Cells.
SLAS discovery : advancing life sciences R & D (2017-03-28)
David M Carter, Edgar Specker, Jessica Przygodda, Martin Neuenschwander, Jens Peter von Kries, Udo Heinemann, Marc Nazaré, Ulrich Gohlke
RÉSUMÉ
Human lysine demethylase (KDM) enzymes (KDM1-7) constitute an emerging class of therapeutic targets, with activities that support growth and development of metastatic disease. By interacting with and co-activating the androgen receptor, the KDM4 subfamily (KDM4A-E) promotes aggressive phenotypes of prostate cancer (PCa). Knockdown of KDM4 expression or inhibition of KDM4 enzyme activity reduces the proliferation of PCa cell lines and highlights inhibition of lysine demethylation as a possible therapeutic method for PCa treatment. To address this possibility, we screened the ChemBioNet small molecule library for inhibitors of the human KDM4E isoform and identified several compounds with IC
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
ML324, ≥98% (HPLC)